Bismuth(III) oxide ≥99%

Supplier: BeanTown Chemical

Synonyms: Bismuth Trioxide, Dibismuth trioxide

143690-1KG 143690-250G 143690-5KG
BT143690-1KGEA 162 USD
BT143690-1KG BT143690-250G BT143690-5KG
Bismuth(III) oxide ≥99%
Bismuth (III) oxide

CAS: 1304-76-3; EC No: 215-134-7; MDL No: MFCD00003462; RTECS: EB2984460 Powder; Molecular Formula: Bi2O3; MW: 465.96


  • RTECS: EB2984460
  • Powder

Formula: BiO₃
MDL Number: MFCD00003462
CAS Number: 1304-76-3
EINECS: 215-134-7

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR