Hydrogen peroxide 29 - 32% (w/w) in water stabilised ACS

Supplier: BeanTown Chemical

141405-4X500ML 141405-500ML 141405-50ML
BT141405-4X500EA 215.99 USD
BT141405-4X500 BT141405-500ML BT141405-50ML
Hydrogen peroxide 29 - 32% (w/w) in water stabilised ACS
Hydrogen peroxide

CAS: 7722-84-1; EC No: 231-765-0; MDL No: MFCD00011333; RTECS: MX0899000 UN No: UN2014; Haz Class: 5.1 (8); Packing Group: II Liquid; Molecular Formula: H2O2; MW: 34.02


  • RTECS: MX0899000
  • Liquid

Formula: H₂O₂
MDL Number: MFCD00011333
CAS Number: 7722-84-1
EINECS: 231-765-0
UN: 2014
ADR: 5.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR