Calcium chloride, anhydrous ≥96.0% ACS

Supplier: BeanTown Chemical

Synonyms: Chloro calcium, Calcium dichloride, CaCl2

140230-250G 140230-50G
BT140230-250GEA 88 USD
BT140230-250G BT140230-50G
Calcium chloride, anhydrous ≥96.0% ACS
Calcium chloride

CAS: 10043-52-4; EC No: 233-140-8; MDL No: MFCD00010903; RTECS: EV9800000 Powder; Molecular Formula: CaCl2; MW: 110.98 Melting Point: 772° Hygroscopic


  • RTECS: EV9800000
  • Hygroscopic
  • Powder

Formula: CaCl₂
Melting Pt: 772 °C
MDL Number: MFCD00010903
CAS Number: 10043-52-4
EINECS: 233-140-8

Order Now

Specification Test Results

Assay 96.0% min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR