Ammonia solution 1.0 N in water volumetric solution

Supplier: BeanTown Chemical

Synonyms: Ammonia aqueous, Ammonium hydroxide solution, Ammonium hydroxide, Ammonia water

136340-1L 136340-4L
BT136340-1LEA 63 USD
BT136340-1L BT136340-4L
Ammonia solution 1.0 N in water volumetric solution
Ammonia solution

CAS: 1336-21-6; EC No: 215-647-6; MDL No: MFCD00066650 Liquid; Linear Formula: NH4OH; MW: 35.05 Density (g/mL): 0.9


  • Liquid

Formula: NH₃
Density: 0.9 g/mL
MDL Number: MFCD00066650
CAS Number: 1336-21-6
EINECS: 215-647-6
UN: 2672
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR