Tin(IV) oxide ≥99.995%

Supplier: BeanTown Chemical

Synonyms: Tin(iv)oxide, Stannic Oxide, Tin dioxide

136065-25G 136065-5G
BT136065-25GEA 460 USD
BT136065-25G BT136065-5G
Tin(IV) oxide ≥99.995%
Tin (IV) oxide

CAS: 18282-10-5; EC No: 242-159-0; MDL No: MFCD00011244; RTECS: XQ4000000 -22 Mesh Powder; Linear Formula: SnO2; MW: 150.71 Melting Point: 1630°; Boiling Point: 1800-1900° Density (g/mL): 6.95; Refractive Index: 1.997


  • RTECS: XQ4000000
  • Refractive index: 1.997
  • -22 Mesh Powder

Formula: O₂Sn
Boiling Pt: 1800…1900 °C
Melting Pt: 1630 °C
Density: 6.95 g/mL
MDL Number: MFCD00011244
CAS Number: 18282-10-5
EINECS: 242-159-0

Order Now

Specification Test Results

Assay 99.995%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR