Trimethyl phosphate ≥98%

Supplier: BeanTown Chemical

Synonyms: Trimethylphosphate

135785-2.5KG 135785-500G
BT135785-2.5KGEA 260 USD
BT135785-2.5KG BT135785-500G
Trimethyl phosphate ≥98%
Trimethyl phosphate

CAS: 512-56-1; EC No: 208-144-8; MDL No: MFCD00008348; RTECS: TC8225000 Liquid; Linear Formula: P(O)(OCH3)3; MW: 140.08 Melting Point: -46°; Boiling Point: 196-198°; Flash point: 150°C (302°F) Density (g/mL): 1.197; Refractive Index: 1.395 Moisture Sensitive


  • RTECS: TC8225000
  • Refractive index: 1.395
  • Moisture Sensitive
  • Liquid

Formula: C₃H₉O₄P
Boiling Pt: 196…198 °C
Melting Pt: –46 °C
Density: 1.197 g/mL
Flash Pt: 150 °C (302 °F)
MDL Number: MFCD00008348
CAS Number: 512-56-1
EINECS: 208-144-8

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR