Mineral oil

Supplier: BeanTown Chemical

135060-1KG
BT135060-1KGEA 56.31 USD
BT135060-1KG
Mineral oil
Mineral oil (8020-83-5)

CAS: 8020-83-5 MDL No: MFCD00131611 Liquid Flash point: 112°C (234°F) Density (g/mL): 0.863; Refractive Index: 1.467


  • Refractive index: 1.467
  • Liquid

Density: 0.863 g/mL
Flash Pt: >112 °C (>234 °F)
MDL Number: MFCD00131611
CAS Number: 8020-83-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR