Iron ≥99.99% (trace metals basis), rods Ø 5 mm

Supplier: BeanTown Chemical

134970-150MM 134970-50MM
BT134970-150MMEA 389 USD
BT134970-150MM BT134970-50MM
Iron ≥99.99% (trace metals basis), rods Ø 5 mm
Iron

CAS: 7439-89-6; EC No: 231-096-4; MDL No: MFCD00010999 Solid; Molecular Formula: Fe; MW: 55.85 Melting Point: 1535°; Boiling Point: 2750° Density (g/mL): 7.86


  • Solid

Formula: Fe
Boiling Pt: 2750 °C
Melting Pt: 1535 °C
Density: 7.86 g/mL
MDL Number: MFCD00010999
CAS Number: 7439-89-6
EINECS: 231-096-4

Order Now

Specification Test Results

Assay 99.99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR