Gold ≥99.999% (trace metals basis), wire Ø 1.0 mm

Supplier: BeanTown Chemical

134960-10CM 134960-2CM 134960-50CM
BT134960-10CMEA 338 USD
BT134960-10CM BT134960-2CM BT134960-50CM
Gold ≥99.999% (trace metals basis), wire Ø 1.0 mm
Gold

CAS: 7440-57-5; EC No: 231-165-9; MDL No: MFCD00003436; RTECS: MD5420000 Solid; Molecular Formula: Au; MW: 196.97 Melting Point: 1063°; Boiling Point: 2808° Density (g/mL): 19.3


  • RTECS: MD5420000
  • Solid

Formula: Au
Boiling Pt: 2808 °C
Melting Pt: 1063 °C
Density: 19.3 g/mL
MDL Number: MFCD00003436
CAS Number: 7440-57-5
EINECS: 231-165-9

Order Now

Specification Test Results

Assay 99.999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR