Ruthenium, Plasma Standard Solution, Ru 1000 µg/ml

Supplier: BeanTown Chemical

134875-500ML
BT134875-100MLEA 167.31 USD
BT134875-100ML BT134875-500ML BT134875-50ML
Ruthenium, Plasma Standard Solution, Ru 1000 µg/ml
Standards Cationic Standards Ruthenium Standards

EC No: 231-595-7.
Haz class: 8
Packing group: II
Signal word: Danger

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR