Imidazole ≥99%

Supplier: BeanTown Chemical

Synonyms: Glyoxaline, 1H-Imidazole, Glyoxalamine

134805-100G 134805-2.5KG 134805-500G
BT134805-100GEA 36.73 USD
BT134805-100G BT134805-2.5KG BT134805-500G
Imidazole ≥99%
Imidazole

CAS: 288-32-4; EC No: 206-019-2; MDL No: MFCD00005183; RTECS: NI3325000 UN No: UN2923; Haz Class: 8 (6.1); Packing Group: III Crystalline; Molecular Formula: C3H4N2; MW: 68.08 Melting Point: 85-92°; Boiling Point: 256°; Flash point: 145°C (293°F)


  • RTECS: NI3325000
  • Crystalline

Formula: C₃H₄N₂
Boiling Pt: 256 °C
Melting Pt: 85…92 °C
Flash Pt: 145 °C (293 °F)
MDL Number: MFCD00005183
CAS Number: 288-32-4
EINECS: 206-019-2
UN: 2923
ADR: 8,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=48eba49e61201d6fffa2bef0c54633a20660c7ef&vid=e7f154e4976b52732539e49bbff60454d5eb82ff&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR