Acetonitrile, anhydrous ≥99.9% animal-free

Supplier: BeanTown Chemical

Synonyms: Methanecarbonitrile, Methane, cyano-, Methyl cyanide, Ethyl nitrile, ACN, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile

128625-4L
BT128625-4LEA 223 USD
BT128625-4L
Acetonitrile, anhydrous ≥99.9% animal-free
Acetonitrile

CAS: 75-05-8; EC No: 200-835-2; MDL No: MFCD00001878; RTECS: AL7700000 UN No: UN1648; Haz Class: 3; Packing Group: II Liquid; Linear Formula: CH3CN; MW: 41.05 Melting Point: -48°; Boiling Point: 81-82°; Flash point: 2°C (36°F) Density (g/mL): 0.786; Refractive Index: 1.344


  • RTECS: AL7700000
  • Refractive index: 1.344
  • Liquid

Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 81…82 °C
Melting Pt: –48 °C
Density: 0.786 g/mL
Flash Pt: 2 °C (36 °F)
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3 II
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.9%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR