Phosphorous acid ≥97%

Supplier: BeanTown Chemical

Synonyms: Phosphonic acid

127760-2KG 127760-500G
BT127760-2KGEA 239.72 USD
BT127760-2KG BT127760-500G
Phosphorous acid ≥97%
Phosphorous acid

CAS: 13598-36-2; EC No: 237-066-7; MDL No: MFCD00137258; RTECS: SZ6400000 UN No: UN2834; Haz Class: 8; Packing Group: III Crystalline; Linear Formula: H3PO3; MW: 82.00 Melting Point: 74°; Boiling Point: 200° (decomposes) Density (g/mL): 1.651 Air Sensitive , Hygroscopic


  • RTECS: SZ6400000
  • Air Sensitive, Hygroscopic
  • Crystalline

Formula: H₃O₃P
Boiling Pt: 200 °C (dec.)
Melting Pt: 74 °C
Density: 1.651 g/mL
MDL Number: MFCD00137258
CAS Number: 13598-36-2
EINECS: 237-066-7
UN: 2834
ADR: 8,III

Order Now

Specification Test Results

Assay 97%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR