N-Acetyl-L-cysteine ≥98%

Supplier: BeanTown Chemical

Synonyms: Acetylcysteine, N-Acetyl-3-mercaptoalanine, NAC, (R)-(+)-N-Acetylcysteine, (R)-2-Acetamido-3-mercaptopropanoic acid, Ac-Cys-OH, N-Acetyl-L-Cysteine, LNAC

124480-100G 124480-25G 124480-500G
BT124480-100GEA 129.57 USD
BT124480-100G BT124480-25G BT124480-500G
N-Acetyl-L-cysteine ≥98%
N-Acetyl-L-cysteine

CAS: 616-91-1; EC No: 210-498-3; MDL No: MFCD00004880; RTECS: HA1660000 Powder; Linear Formula: HSCH2CH(NHCOCH3)COOH; Molecular Formula: C5H9NO3S; MW: 163.20 Melting Point: 106-108°


  • RTECS: HA1660000
  • Powder

Formula: C₅H₉NO₃S
Melting Pt: 106…108 °C
MDL Number: MFCD00004880
CAS Number: 616-91-1
EINECS: 210-498-3

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR