Tetramethylthiuram monosulfide ≥97%

Supplier: BeanTown Chemical

Synonyms: Bis(dimethylthiocarbamyl) sulphide, Bis(dimethylthiocarbamyl) sulfide

123955-100G 123955-25G 123955-500G
BT123955-100GEA 64 USD
BT123955-100G BT123955-25G BT123955-500G
Tetramethylthiuram monosulfide ≥97%
Tetramethylthiuram monosulphide

CAS: 97-74-5; EC No: 202-605-7; MDL No: MFCD00014870; RTECS: WQ1750000 Powder; Linear Formula: [(CH3)2NCS]2S; MW: 208.37 Melting Point: 107-111°; Flash point: 156°C (313°F) Density (g/mL): 1.37


  • RTECS: WQ1750000
  • Powder

Formula: C₆H₁₂N₂S₃
Melting Pt: 107…111 °C
Density: 1.37 g/mL
Flash Pt: 156 °C (313 °F)
MDL Number: MFCD00014870
CAS Number: 97-74-5
EINECS: 202-605-7
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Assay 97%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR