Maleic hydrazide ≥97%

Supplier: BeanTown Chemical

Synonyms: 1,2-Dihydro-3,6-pyridazinedione, 3,6-Dihydroxypyridazine, 3,6-Pyridazinediol, Maleic acid Hydrazide

123810-100G 123810-500G
BT123810-100GEA 46 USD
BT123810-100G BT123810-500G
Maleic hydrazide ≥97%
Maleic hydrazide

CAS: 123-33-1; EC No: 204-619-9; MDL No: MFCD00006665; RTECS: UR5950000 Powder; Molecular Formula: C4H4N2O2 ; MW: 112.09 Melting Point: 299-301° (decomposes); Flash point: 300°C (572°F)


  • RTECS: UR5950000
  • Powder

Formula: C₄H₄N₂O₂
Melting Pt: 299…301 °C (dec.)
Flash Pt: 300 °C (572 °F)
MDL Number: MFCD00006665
CAS Number: 123-33-1
EINECS: 204-619-9

Order Now

Specification Test Results

Assay 97%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR