Nitric acid 65 - 70% ≥99.999% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Nitric acid solution

123485-100ML 123485-500ML
BT123485-100MLEA 58.57 USD
BT123485-100ML BT123485-500ML
Nitric acid 65 - 70% ≥99.999% (trace metals basis)
Nitric acid

CAS: 7697-37-2; EC No: 231-714-2; MDL No: MFCD00011349; RTECS: QU5775000 UN No: UN2031; Haz Class: 8 (5.1); Packing Group: II Liquid; Molecular Formula: HNO3; MW: 63.01 Boiling Point: 120.5° Density (g/mL): 1.413


  • RTECS: QU5775000
  • Liquid

Formula: HNO₃
MW: 63.01
Boiling Pt: 87
Melting Pt: -42
Density: 1.413 g/mL
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8 II
Merck Index: 13,06610

Order Now

Specification Test Results

Assay 99.999%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR