Calcium pantothenate ≥98%

Supplier: BeanTown Chemical

Synonyms: Calcium D-Pantothenate, D-Pantothenic acid calcium salt, D-Calcium pantothenate, Calcium (R)-3-(2,4-dihydroxy-3,3-dimethylbutanamido)propanoate, Vitamin B5 calcium salt, Calcium D-(+)-pantothenate

121400-100G 121400-500G
BT121400-100GEA 70.01 USD
BT121400-100G BT121400-500G
Calcium pantothenate ≥98%
Calcium pantothenate

CAS: 137-08-6; EC No: 205-278-9; MDL No: MFCD00002766; RTECS: RU4375000 Crystalline/Powder; Molecular Formula: C18H32CaN2O10; MW: 476.54 Hygroscopic


  • RTECS: RU4375000
  • Hygroscopic
  • Crystalline/Powder

Formula: C₁₈H₃₂CaN₂O₁₀
MDL Number: MFCD00002766
CAS Number: 137-08-6
EINECS: 205-278-9

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR