Hafnium, Plasma Standard Solution, Hf 10000 µg/ml

Supplier: BeanTown Chemical

121275-500ML
BT121275-100MLEA 187.29 USD
BT121275-100ML BT121275-500ML BT121275-50ML
Hafnium, Plasma Standard Solution, Hf 10000 µg/ml
Standards Cationic Standards Hafnium Standards

EC No: 231-595-7
Haz class: 8
GHS symbol: GHS05
Packing group: III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR