Avanti™ J-15 Series Benchtop Centrifuges, Beckman Coulter®
Supplier: Beckman Coulter
B99516 C01994 C01995 B99517 C19391 C19392 C19393 C19394 C19397 C19398 C19399 C19400 C19403 C19404 C19405 C19406 C19409 C19410 C19411 C19412 C19415 C19416 C19417 C19418 C19421 C19422 C19423 C19424
76102-904EA 15473.4 USD
76102-904 76102-920 76102-922 76102-924 BKC19391 BKC19392 BKC19393 BKC19394 BKC19397 BKC19398 BKC19399 BKC19400 BKC19403 BKC19404 BKC19405 BKC19406 BKC19409 BKC19410 BKC19411 BKC19412 BKC19415 BKC19416 BKC19417 BKC19418 BKC19421 BKC19422 BKC19423 BKC19424
Avanti™ J-15 Series Benchtop Centrifuges, Beckman Coulter®
Centrifuges Benchtop Centrifuges
The Avanti™ J-15 series of benchtop centrifuges (refrigerated or ventilated) leverage the ultra harmonic technology, designed to protect your sample and increase workflow time efficiencies.
  • Ultra harmonic technology
  • Workflow optimization
  • Sample protection
  • Time efficiency
  • Multi-dimension imbalance tolerance
The Avanti™ J-15 series of benchtop centrifuges is part of a new family of life science equipment that provides the exceptional performance.
Refrigerated (Avanti™ J-15R) or ventilated (Avanti™ J-15), these centrifuges use ultra harmonicMore Product Information