HarvestLine System Liners, Beckman Coulter
Supplier: Beckman Coulter
358299 369264 361690 369256 C15216 C15217 C15218 C19681
BK358299PK 203.62 USD
BK358299 BK369264 BK361690 BK369256 76292-060 76292-062 76292-064 76292-066 76292-128 76292-130 76292-132 76292-134 76292-136 76294-184 76294-186
HarvestLine System Liners, Beckman Coulter
Bags Media Bags
Faster, safer, more efficient harvesting and now it's all in the bag. Minimize non-value-added tasks e.g. bottle cleaning, autoclaving and scraping. Reduce exposure to potentially harmful biologics.
  • Prevent contamination
  • Reduce repetitive motions
  • Prevent liquid spillage
  • Reduce needed storage space
  • Achieve maximum ample yield
  • Expedite freeze/thaw lysing
Safe, efficient workflows are as critical to study as quality samples and to harvest high-quality samples in fewer steps from prep to cleanup.
Optional sterile bottle liners, optimized speed, safetyMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable