2-Propanol, low water content ≥99.90%

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Isopropyl rubbing alcohol, sec-Propanol, Isopropanol, 2-Hydroxypropane, Isopropyl Rubbing Alcohol, 2-Hydroxy propane, IPA, sec-Propyl Alcohol, Dimethyl carbinol, Propane, 2-hydroxy-, Isopropyl alcohol, Propan-2-ol

SR324AC-204L
BJSR324AC-204LEA 5868.12 USD
BJSR324AC-204L
2-Propanol, low water content ≥99.90%
2-Propanol

Isopropyl Alcohol low water content

Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82
Melting Pt: -89
Density: 0.784…0.786 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3 II
Merck Index: 13,05228

Order Now

Specification Test Results

gC Analysis 99.9 % min.
refractive Index (20ºC) 1.3766 - 1.3778
residue 1 mg/L max.
uV Absorbance @ 225 nm 0.16 AU max.
uV Absorbance @ 250 nm 0.02 AU max.
uV Absorbance @ 300 nm 0.005 AU max.
uV Absorbance @ 400 nm 0.005 AU max.
uV Cutoff 205 nm max.
water by Karl Fischer Titration 0.01 % max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR