Ethyl acetate ≥99.5%, B&J Brand™ for HPLC, for GC (residue analysis), for spectrophotometry, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Acetic acid ethyl ester, Acetic ether, Acetoxyethane, Acetidin, Ethyl acetic ester, Ethyl ethanoate, EtOAc, Vinegar naphtha, Acetic acid ethyl ester

NS100-20
BJNS100-20EA 1425.89 USD
BJNS100-20
Ethyl acetate ≥99.5%, B&J Brand™ for HPLC, for GC (residue analysis), for spectrophotometry, Burdick & Jackson™
Ethyl acetate

  • Minimal UV contaminants
  • High GC purity
  • Low water content
  • Outstanding lot-to-lot consistency


UNSPSC Code: 12191602

Formula: CH₃COOC₂H₅
MW: 88.11 g/mol
Boiling Pt: 77.1
Melting Pt: -83
Density: 0.9006 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
UN: 1173
ADR: 3 II
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Electron Capture GC 10 ng/L max.
Water by Karl Fischer Titration 0.03 % max.
GC Analysis 99.9 % min.
Refractive Index (20ºC) 1.3719 - 1.3727
Residue 1 mg/L max.
UV Absorbance @ 275 nm 0.1 AU max.
UV Absorbance @ 300 nm 0.01 AU max.
UV Absorbance @ 325 nm 0.005 AU max.
UV Absorbance @ 350 nm 0.005 AU max.
UV Cutoff 256 nm max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR