Glycerine 99,0 ACS organic_synthesis_LC_prep, General Laboratory Use

Supplier: Honeywell Research Chemicals

Synonyms: Glyceritol, Glysanin, 1,2,3-Propanetriol, Osmoglyn, Glycerin, Glycyl alcohol, Glyrol, Propane-1,2,3-triol, Propanetriol, Trihydroxypropane, Glycerol

LP380-4P
BJLP380-4PCS 1531.52 USD
BJLP380-4P
Glycerine 99,0 ACS organic_synthesis_LC_prep, General Laboratory Use
Glycerine

ACS GENERAL USE SOLVENTS


  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Formula: HOCH₂CH(OH)CH₂OH
MW: 92.09 g/mol
Boiling Pt: 290
Melting Pt: 18.6
Density: 1.261 g/cm³ (20 °C)
Flash Pt: 199 °C
Storage Temperature: Ambient
MDL Number: MFCD00004722
CAS Number: 56-81-5
Merck Index: 13,04497
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=86a15453b54e7106c121d32e4e716385003c4507&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99.7 % min.
Water by Karl Fischer Titration 0.3 % max.
Chlorides 10 ppm max.
Chlorinated Compounds (as Cl) 30 ppm max.
Color (APHA) 20 max.
Fatty Acids and Esters Passes test
Heavy Metals (as Pb) 5 ppm max.
Residue after Ignition 0.01 % max.
Specific Gravity (25ºC/25ºC) 1.2613 min.
Sulfates 20 ppm max.

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR