Tetrahydrofuran ≥99.0% stabilised ACS analytical reagent, general laboratory use

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Oxolane, 1,4-epoxy-Butane, Butylene oxide, Butane alpha,delta-oxide, Tetramethylene oxide, THF, Oxacyclopentane, Cyclotetramethylene oxide, Diethylene oxide

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)

LP341-4 LP341-19
BJLP341-4CS 2266.57 USD
BJLP341-4 BJLP341D19
Tetrahydrofuran ≥99.0% stabilised ACS analytical reagent, general laboratory use
Tetrahydrofuran

ACS GENERAL USE SOLVENTS


  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications

Formula: C₄H₈O
Boiling Pt: 11.8 %(V)
Melting Pt: –108.5 °C
Density: 0.8892 g/cm³ (20 °C)
Flash Pt: –14 °C
Storage Temperature: Ambient
MDL Number: MFCD00005356
CAS Number: 109-99-9
EINECS: 203-726-8
UN: 2056
ADR: 3,II
REACH: 05-2114615188-48
Merck Index: 12,09351
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b2d7611caf0127fbef941994edd1d7e69487d54a&vid=2a64e21bd2e13636c3038adf839b3e9ff18509c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Passes test
Assay by GC Analysis 99 % min.
Color (APHA) 20 max.
Peroxides (as H₂O₂) at time of packaging 0.015 % max.
Residue after Evaporation 0.03 % max.
Water by Karl Fischer Titration 0.05 % max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR