Formic Acid/Acetonitrile Blend, LabReady™ LC-MS, Solvent Blend for LC-MS, for HPLC, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger

LC441-1 LC441-2.5 LC441-4X4L
BJLC441-1CS 1884 USD
BJLC441-1 BJLC441-2.5 BJLC441-4L
Formic Acid/Acetonitrile Blend, LabReady™ LC-MS, Solvent Blend for LC-MS, for HPLC, Burdick & Jackson™
Acetonitrile with 0,1% (v/v) Formic acid

LabReady™ Solvent Blends


  • Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
  • Honeywell Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
  • Honeywell LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
  • Honeywell’s high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk

Boiling Pt: 82 °C (1.013 hPa)
Melting Pt: –46 °C
Density: 0.7767 g/cm³ (25 °C)
Flash Pt: 8 °C
Storage Temperature: Ambient
EINECS: 200-579-1
UN: 1648
ADR: 3 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR