Trichloroacetic acid (TCA) 3% (w/v) in dichloromethane, BioSyn™ detritylation reagent, for DNA/RNA synthesis, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: TCA, Trichloroethanoic acid, Aceto-Caustin, TCA

BR605-2 BR605-4 BR605-20 BR605-4X2.5L
BJBR605-2CS 1749.78 USD
BJBR605-2 BJBR605-4 BJBR605-20 BJBR605-4X2.5L
Trichloroacetic acid (TCA) 3% (w/v) in dichloromethane, BioSyn™ detritylation reagent, for DNA/RNA synthesis, Burdick & Jackson™
Trichloroacetic acid (TCA)

Burdick & Jackson® BioSyn® reagents & solvents for DNA/RNA synthesis


  • Honeywell Burdick & Jackson® BioSyn® Solvents and Reagents are specifically processed and purified to exacting specifications, ensuring outstanding synthesis efficiency
  • ideal for oligonucleotide synthesis regardless of the application
  • All solvents and reagents are filtered (0.2-micron for oxidation reagents and 0.1-micron for all other reagents) to minimize particulate contamination


UNSPSC Code: 12350000

Formula: Cl₃CCOOH
Boiling Pt: 40 °C
Melting Pt: –95 °C
Density: 1.33 g/cm³ (20 °C)
Flash Pt: Does not flash (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00004177
CAS Number: 76-03-9
UN: 2922
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ae34130684af3f73eccad44eac686e325855232d&vid=b600bf2ba67ce275263e6172675e131d3f263837&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR