Silver nitrate 0.1 M AgNO3 (0.1N) volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Warning

Synonyms: Lunar caustic, Silver mononitrate, Nitric acid silver salt

72271-1L
BJ72271-1LEA 238 USD
BJ72271-1L
Silver nitrate 0.1 M AgNO3 (0.1N) volumetric solution, Fluka™
Silver nitrate
Formula: AgNO₃
MW: 169.87 g/mol
Boiling Pt: 444
Melting Pt: 210
Density: 1.010 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003414
CAS Number: 7761-88-8
UN: 3264
ADR: 8 III
Merck Index: 12,08661
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f56d5ff35f4c0937041d9629e42aae69bf1987b3&vid=7ca52a55b378bddc27fb4a51ace3ae27a0451c75&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 0.0998 - 0.1002 mol/l
Calibrated Against sodium chloride


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR