N-Methyl-2-pyrrolidone (NMP) ≥99.0% ACS, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: 1-Methyl-2-pyrrolidinone, N-Methyl-2-pyrrolidinone,NMP, N-Methylpyrrolidinone, N-Methyl-2-pyrrolidinone, N-Methyl-alpha-pyrrolidone, M-Pyrol, N-methylpyrrolidone, N-Methyl-gamma-butyrolactam, 1-Methyl-2-pyrrolidone, NMP, N-Methylpyrrolidone, N-Methyl-2-pyrrolidone, Methyl Pyrrolidone

443778-1L 443778-2.5L
BJ443778-1LEA 177.02 USD
BJ443778-1L BJ443778-2.5L BJ443778-20L
N-Methyl-2-pyrrolidone (NMP) ≥99.0% ACS, Riedel-de Haën™
N-Methyl-2-pyrrolidone (NMP)

ACS GENERAL USE SOLVENTS


  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications


Beilstein Number: 106420

Formula: C₅H₉NO
MW: 99.13 g/mol
Boiling Pt: 202
Melting Pt: -24
Density: 1.028 g/cm³ (20 °C)
Flash Pt: 9.5 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00003193
CAS Number: 872-50-4
EINECS: 212-828-1
Merck Index: 13,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=32d156c242c81ef2974339109ced3895beda87fd&vid=06c87bbb3b5050362f47a8fc836cedc8a6d51530&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.0 %
Water (Karl Fischer) Max. 0.05 %
Free amine Max. 0.01 %
Chloride (Cl) Max. 0.0001 %
APHA Max. 50
IR spectrum complying
Vapor Density 3.4 (vs air)
Vapor Pressure 0.99 mmHg ( 40 °C)
Autoignition Temperature 518 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR