Sodium hydroxide concentrate for dilution to 2L, 1 M NaOH (1N) standard volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

38212-1EA
BJ38212-1EAEA 119.51 USD
BJ38212-1EA
Sodium hydroxide concentrate for dilution to 2L, 1 M NaOH (1N) standard volumetric solution, Fluka™
Sodium hydroxide

concentrate for 2L standard solution

Formula: NaOH
Boiling Pt: 120 °C
Melting Pt: 319 °C
Density: 1.350 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Factor 0.998 - 1.002
BAM/EMPA reference material effective
BAM/EMPA Certificate Number effective
Traceable to NIST SRM effective


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR