1,2-Diaminocyclohexanetetraacetic acid disodium salt 0.1 M DCTA-Na4 aqueous solution Reag. Ph. Eur. Volumetric, Complexometry, Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: CDTA disodium salt

35103-6X1L
BJ35103-1LCS 1280.86 USD
BJ35103-1L BJ35103-6X1L
1,2-Diaminocyclohexanetetraacetic acid disodium salt 0.1 M DCTA-Na4 aqueous solution Reag. Ph. Eur. Volumetric, Complexometry, Fluka™
1,2-Diaminocyclohexanetetraacetic acid disodium salt
Formula: C₁₄H₂₀N₂Na₂O₈
MW: 390.3 g/mol
CAS Number: 5786-78-7

Order Now

Specification Test Results

Factor 0.999 - 1.001

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR