n-Heptane fraction 99 isomer_n_GC, Puriss. p.a. Reag. Ph. Eur.

Supplier: Honeywell Research Chemicals
Danger

Synonyms: high purity solvents

32004-2.5L
BJ32004-2.5LEA 161.04 USD
BJ32004-2.5L
n-Heptane fraction 99 isomer_n_GC, Puriss. p.a. Reag. Ph. Eur.
n-Heptane
Formula: H₃C(CH₂)₅CH₃
MW: 100.20 g/mol
Boiling Pt: 98.4
Melting Pt: -90.6
Density: 0.69 g/cm³ (15 °C)
Flash Pt: 6.7 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00009544
CAS Number: 142-82-5
EINECS: 205-563-8
UN: 1206
ADR: 3 II
Merck Index: 13,04679
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8ff97126664c92362bf0aa2645719548c99f0533&vid=6722523a326fdf1ebabd08a2b8656dd57300787f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay n-Heptane (GC) Min. 99 %
Boiling range 97 - 98 °C
Density (D 20/4) 0.683 - 0.685
Refractive index (n 20/D) 1.3870 - 1.3880
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.01 %
Free acid (as CH₃COOH) Max. 0.0005 %
Aluminium (Al) Max. 0.00005 %
Boron (B) Max. 0.000002 %
Barium (Ba) Max. 0.00001 %
Calcium (Ca) Max. 0.00005 %
Cadmium (Cd) Max. 0.000005 %
Cobalt (Co) Max. 0.000002 %
Chromium (Cr) Max. 0.000002 %
Copper (Cu) Max. 0.000002 %
Iron (Fe) Max. 0.00001 %
Magnesium (Mg) Max. 0.00001 %
Manganese (Mn) Max. 0.000002 %
Nickel (Ni) Max. 0.000002 %
Lead (Pb) Max. 0.00001 %
Tin (Sn) Max. 0.00001 %
Zinc (Zn) Max. 0.00001 %
S-compounds (as S) Max. 0.005 %
Reaction against H₂SO₄ complying


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR