Ethyl acetate ≥99.5% ACS, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Acetic acid ethyl ester, Acetic ether, Acetoxyethane, Acetidin, Ethyl acetic ester, Ethyl ethanoate, EtOAc, Vinegar naphtha, Acetic acid ethyl ester

319902-500ML 319902-6X500ML
BJ319902-500MLEA 109.26 USD
BJ319902-500ML BJ319902-6X1L BJ3199026500ML
Ethyl acetate ≥99.5% ACS, Riedel-de Haën™
Ethyl acetate

ACS GENERAL USE SOLVENTS


  • value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • variations and decrease issues traceable to solvent contamination
  • all solvents are tested to meet ACS general use specifications


Beilstein Number: 506104
RTECS Number: AH5425000
UNSPSC Code: 12191602

Formula: CH₃COOC₂H₅
Boiling Pt: 77 °C (1.013 hPa)
Melting Pt: –84 °C
Density: 0.901 g/cm³
Flash Pt: 11.5 %(V)
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identity (IR) complying
Assay (GC) Min. 99.50 %
Non-volatile matter Max. 0.003 %
Water (Karl Fischer) Max. 0.2 %
Titrable Acid Max. 0.0009 meq/g
Reaction against H₂SO₄ complying
APHA Max. 10
Vapor Pressure 73 mmHg ( 20 °C)
Autoignition Temperature 801 °F
Vapor Density 3 (20 °C, vs air)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR