Chromium(III) nitrate nonahydrate ≥98.0%, Puriss. p.a., Fluka™

Supplier: Honeywell Research Chemicals
Warning

Synonyms: Chromium trinitrate nonahydrate, Chromic nitrate nonahydrate, Chromium nitrate nonahydrate

31313-25KG 31313-500G
BJ31313-25KGEA 7240.51 USD
BJ31313-25KG BJ31313-500G
Chromium(III) nitrate nonahydrate ≥98.0%, Puriss. p.a., Fluka™
Chromium (III) nitrate nonahydrate
Formula: CrN₃O₉·9H₂O
Boiling Pt: Decomposes on heating
Melting Pt: 54.8 °C
Density: 1.89 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149659
CAS Number: 7789-02-8
UN: 3260
ADR: 8,III

Order Now

Specification Test Results

Assay 98.0 - 102.0 %
Calcium (Ca) Max. 50 ppm
Cadmium (Cd) Max. 10 ppm
Copper (Cu) Max. 10 ppm
Iron (Fe) Max. 200 ppm
Potassium (K) Max. 50 ppm
Sodium (Na) Max. 50 ppm
Nickel (Ni) Max. 50 ppm
Lead (Pb) Max. 20 ppm
Zinc (Zn) Max. 10 ppm
Total S (as SO₄) Max. 50 ppm
Chloride (Cl) Max. 50 ppm
Ammonium (NH₄) Max. 10 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR