Petroleum spirit, 60…80 °C, Spectrophotometric Grade, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Gasoline, Petroleum ether, Petrol, Benzine

261734-1L
BJ261734-1LEA 143.95 USD
BJ261734-1L
Petroleum spirit, 60…80 °C, Spectrophotometric Grade, Riedel-de Haën™
Petroleum spirit

  • Spectrophotometric grade solvents are high purity solvents made specifically for applications using spectrophotometers.


RTECS Number: OI6180000
UNSPSC Code: 12191502

Boiling Pt: 40…60 °C (1.013 hPa)
Density: 0.65 g/cm³ (20 °C)
Flash Pt: –30 °C
Storage Temperature: Ambient
MDL Number: MFCD00081849
CAS Number: 64742-49-0
UN: 3295
ADR: 3 II

Order Now

Specification Test Results

Explosion Limit 8 %
Identity (IR) complying
Non-volatile matter Max. 0.0005 %
Water (Karl Fischer) Max. 0.02 %
Absorbance at 400 nm Max. 0.01
Absorbance at 260 nm Max. 0.01
Absorbance at 240 nm Max. 0.05
Absorbance at 230 nm Max. 0.10
Absorbance at 220 nm Max. 0.30
Absorbance at 210 nm Max. 1.0
Vapor Density 2.5 (vs air)
Refractive Index n20/D 1.363 (lit.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR