Diethyl ether ≥99.5% (by GC) stabilised, Puriss., meets analytical specification of BP, Ph. Eur., Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane, Ethyl ether

Stabilization: Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 5 - 8 ppm

24004-25L 24004-135KG 24004-1L 24004-2.5L 24004-4X2.5L 24004-4X5L 24004-5L 24004-6X1L
BJ24004-25LEA 1756.23 USD
BJ24004-25L BJ240042.5LALU BJ24004-135KG BJ24004-1L BJ24004-2.5L BJ24004-4X2.5L BJ24004-4X5L BJ24004-5L BJ24004-6X1L
Diethyl ether ≥99.5% (by GC) stabilised, Puriss., meets analytical specification of BP, Ph. Eur., Riedel-de Haën™
Diethyl ether

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.


Beilstein Number: 1696894
RTECS Number: KI5775000
UNSPSC Code: 12191602

Formula: (CH₃CH₂)₂O
Boiling Pt: 34…35 °C (1.013 hPa)
Melting Pt: –116 °C (1.013 hPa)
Density: 0.71 g/cm³ (20 °C)
Flash Pt: –45 °C (1.013 hPa)
Storage Temperature: Ambient
MDL Number: MFCD00011646
CAS Number: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=3dd6c545e58e36e1004e828026523fb0e3bcbbcd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Refractive Index n20/D 1.3530 (lit.)
Identity A,B complying
Assay (GC) Min. 99.5 %
Boiling range 34,0 - 35,0 °C
Density (D 20/20) 0.714 - 0.716
Non-volatile matter Max. 0.002 %
Water (Karl Fischer) Max. 0.2 %
Free acid (as CH₃COOH) Max. 0.002 %
Aldehydes complying
Peroxides (as H₂O₂) Max. 0.0002 %
Acidity complying
Assay BHT (GC) 5 - 8 ppm
Residual solvents complying
Vapor Density 2.6 (vs air)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR