Iodine ≥99.8%, solid ACS, Fluka™

Supplier: Honeywell Research Chemicals
Danger

207772-100G 207772-1KG 207772-2.5KG
BJ207772-100GEA 140 USD
BJ207772-100G BJ207772-1KG BJ207772-2.5KG
Iodine ≥99.8%, solid ACS, Fluka™
Iodine

Honeywell ACS solvents for general laboratory use are:


  • A value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
  • suitable for less demanding applications such as routine isocratic HPLC
  • Variations and decrease issues traceable to solvent contamination
  • All solvents are tested tmeet ACS general use specifications

Formula: I₂
MW: 253.81 g/mol
Boiling Pt: 184.3
Melting Pt: 114
Density: 4.930 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011355
CAS Number: 7553-56-2
EINECS: 231-442-4
UN: 3495
ADR: 8 III
Merck Index: 13,05036

Order Now

Specification Test Results

Assay Min. 99.8 %
Non-volatile matter Max. 0.01 %
Bromine and chlorine (as chloride) Max. 0.005 %
Form Solid
Vapor Pressure 0.31 mmHg ( 25 °C)
Resistivity 1.3E15 μΩ-cm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR