2-Propanol ≥99.9% (by GC), TraceSELECT™ for metal speciation analysis, Riedel-de Haën™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Isopropyl rubbing alcohol, sec-Propanol, Isopropanol, 2-Hydroxypropane, Isopropyl Rubbing Alcohol, 2-Hydroxy propane, IPA, sec-Propyl Alcohol, Dimethyl carbinol, Propane, 2-hydroxy-, Isopropyl alcohol, Propan-2-ol

04516-1L
BJ04516-1LEA 156.35 USD
BJ04516-1L
2-Propanol ≥99.9% (by GC), TraceSELECT™ for metal speciation analysis, Riedel-de Haën™
2-Propanol

High purity solvents for speciation analysis by LC-ICP-MS. Rigorous purification procedures followed by UV spectroscopy, IC, and ICP-MS testing to assure high chemical purity and high UV transmittance. The blank values for metal traces in these solvents are in the ppb range or lower.

Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82
Melting Pt: -89
Density: 0.784…0.786 g/cm³ (20 °C)
Flash Pt: 12 °C
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1219
ADR: 3 II
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) Min. 99.9 %
Water (Karl Fischer) Max. 0.05 %
Silver (Ag) Max. 0.5 ppb
Aluminium (Al) Max. 5 ppb
Arsenic (As) Max. 1 ppb
Gold (Au) Max. 0.5 ppb
Barium (Ba) Max. 0.5 ppb
Beryllium (Be) Max. 0.5 ppb
Bismuth (Bi) Max. 0.5 ppb
Calcium (Ca) Max. 30 ppb
Cadmium (Cd) Max. 0.5 ppb
Cerium (Ce) Max. 0.5 ppb
Cobalt (Co) Max. 0.5 ppb
Chromium (Cr) Max. 1 ppb
Cesium (Cs) Max. 0.5 ppb
Copper (Cu) Max. 1 ppb
Iron (Fe) Max. 5 ppb
Gallium (Ga) Max. 0.5 ppb
Mercury (Hg) Max. 1 ppb
Indium (In) Max. 0.5 ppb
Iridium (Ir) Max. 0.5 ppb
Potassium (K) Max. 10 ppb
Lithium (Li) Max. 0.5 ppb
Magnesium (Mg) Max. 10 ppb
Manganese (Mn) Max. 5 ppb
Molybdenum (Mo) Max. 0.5 ppb
Sodium (Na) Max. 250 ppb
Nickel (Ni) Max. 5 ppb
Lead (Pb) Max. 0.5 ppb
Palladium (Pd) Max. 1 ppb
Platinum (Pt) Max. 0.5 ppb
Rubidium (Rb) Max. 0.5 ppb
Rhodium (Rh) Max. 0.5 ppb
Ruthenium (Ru) Max. 0.5 ppb
Antimony (Sb) Max. 0.5 ppb
Selenium (Se) Max. 1 ppb
Tin (Sn) Max. 0.5 ppb
Strontium (Sr) Max. 0.5 ppb
Titanium (Ti) Max. 5 ppb
Thallium (Tl) Max. 0.5 ppb
Uran (U) Max. 0.5 ppb
Vanadium (V) Max. 0.5 ppb
Zinc (Zn) Max. 5 ppb
Zirconium (Zr) Max. 0.5 ppb
Bromide (Br) Max. 10 ppb
Chloride (Cl) Max. 25 ppb
Fluoride (F) Max. 10 ppb
Iodide (I) Max. 10 ppb
Nitrite (NO₂) Max. 10 ppb
Nitrate (NO₃) Max. 25 ppb
Phosphate (PO₄) Max. 50 ppb
Sulfate (SO₄) Max. 50 ppb
Peroxides (as H₂O₂) Max. 5 ppm
Propionaldehyde (GC) Max. 20 ppm
Ketones (as CH₃COCH₃) Max. 20 ppm
Transmittance from 260 nm Min. 98 %
Transmittance at 230 nm Min. 75 %
Transmittance at 220 nm Min. 50 %
Autoignition Temperature 750 °F


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR