Ethylene glycol, Technical Grade suitable as anti-freezing agent (with corrosion-inhibitor), blue

Supplier: Honeywell Research Chemicals
Warning

Synonyms: Monoethylene glycol, Ethane-1,2-diol, 2-Hydroxyethanol, Ethylene alcohol, Ethanediol, Glycol alcohol, 1,2-Dihydroxy ethane, 1,2-Ethanediol, 1,2-Dihydroxyethane, Glycol, Dihydroxyethane

03770-5L
BJ03770-1LEA 169 USD
BJ03770-1L BJ03770-5L
Ethylene glycol, Technical Grade suitable as anti-freezing agent (with corrosion-inhibitor), blue
Ethylene glycol

  • Chemicals that do not have an established standard set for quality and impurity levels or for products where the purity is <90%
  • Variation in color and physical form is possible

Formula: HOCH₂CH₂OH
MW: 62.07 g/mol
Melting Pt: -13
Density: 1.115 g/cm³
Flash Pt: 111 °C
Storage Temperature: Ambient
MDL Number: MFCD00002885
CAS Number: 107-21-1
Merck Index: 13,03832
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4d1c97392851ba184f445f6ee912f84631f98d9&vid=2002300f174929a030d66d9f1394eb6bee89b7dc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Refractive Index n20/D 1.431 (lit.)
Appearance (color) complying
Identity (IR) complying
Refractive index (n 20/D) 1.428 - 1.438
Water (Karl Fischer) 3 - 7 %
Vapor Pressure 0.08 mmHg ( 20 °C)
Vapor Density 2.1 (vs air)
Explosion Limit 15.3 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR