Acetonitrile ≥99.9%, B&J Brand™, gradient grade for HPLC, for pesticide residue analysis, for spectrophotometry, Burdick & Jackson™

Supplier: Honeywell Research Chemicals
Danger

Synonyms: Methanecarbonitrile, Methane, cyano-, Methyl cyanide, Ethyl nitrile, ACN, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile

015-1L NP015-20 015-4 NS015-20
BJ015-1LCS 1743 USD
BJ015-1L BJNP015-20 BJ015-4 BJNS015-20
Acetonitrile ≥99.9%, B&J Brand™, gradient grade for HPLC, for pesticide residue analysis, for spectrophotometry, Burdick & Jackson™
Acetonitrile

For HPLC, liquid and gas chromatography, pesticide residue analysis, spectophotometry, organic synthesis and combinatorial chemistry.


  • Minimal UV contaminants
  • High GC purity
  • Low water content
  • Outstanding lot-to-lot consistency

Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 82 °C (1.013 hPa)
Melting Pt: –46 °C
Density: 0.7822 g/cm³ (20 °C)
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3 II
REACH: 01-2119471307-38
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Electron Capture GC 10 ng/L max.
Fluorescence (as quinine base) 0.3 ppb max.
GC Analysis 99.9 % min.
HPLC Gradient @ 215 nm 0.005 AU max.
HPLC Gradient @ 254 nm 0.001 AU max.
Refractive Index (20ºC) 1.3434 - 1.3446
Residue 1 mg/L max.
UV Cutoff 190 nm max.
Water by Karl Fischer Titration 0.01 % max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR