Sodium thiosulfate 0.05 M Na2S2O3 (0.05N) volumetric solution, Fluka™

Supplier: Honeywell Research Chemicals

Synonyms: di-Sodium thiosulfate, Sodiumthiosulphate, Sodium hyposulphite, Antichlor, di-Sodium thiosulphate, Sodiumthiosulfate, Sodium hyposulfite

34449-10L-VP 34449-1L
BJ34449-10L-VPEA 641.45 USD
BJ34449-10L-VP BJ-34449-1L
Sodium thiosulfate 0.05 M Na2S2O3 (0.05N) volumetric solution, Fluka™
Sodium thiosulphate
Formula: Na₂S₂O₃
MW: 158.11 g/mol
Boiling Pt: 100
Melting Pt: 48
Density: 1.0 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00003499
CAS Number: 7772-98-7
EINECS: 231-791-2
Merck Index: 12,08844

Order Now

Specification Test Results

Factor 0.999 - 1.001


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR