Sodium hydroxide (1.3 - 12 mol/l; 1.3 - 12 N) in aqueous solution 40% (w/w) in aqueous solution analytical reagent, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Caustic soda, Caustic soda, Sodium hydrate, Lye, Soda lye

BDH7994-4 BDH7994-6 BDH7994-7
BDH7994-4CS 383.1 USD
BDH7994-4 BDH7994-6 BDH7994-7
Sodium hydroxide (1.3 - 12 mol/l; 1.3 - 12 N) in aqueous solution 40% (w/w) in aqueous solution analytical reagent, VWR Chemicals BDH®
Sodium hydroxide

Made with deionized water.

Formula: NaOH
Boiling Pt: 109 °C (1013 hPa)
Density: ≤1.11 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1824
ADR: 8,II

Order Now

Specification Test Results

Appearance/color Clear liquid
Shelf-life 3 years
Assay 39.60 - 40.40 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR