VWR® Sodium Phosphate 0.1 M, pH 7.0 Buffer
Supplier: VWR International
BDH7895-4
BDH7895-4CS 680.27 USD
BDH7895-4
VWR® Sodium Phosphate 0.1 M, pH 7.0 Buffer
Buffers Cell Culture Buffers
Made with sodium phosphate monobasic monohydrate, sodium hydroxide solution and deionized water.