Acetone ACS for production, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: 2-Propanone, DMK, Propanone, Dimethyl ketone

BDH2002-55GLM BDH2002-5GLMS BDH2002-1GLP
BDH2002-55GLMEA 4300.72 USD
BDH2002-55GLM BDH2002-5GLMS BDH2002-1GLP
Acetone ACS for production, VWR Chemicals BDH®
Acetone

Clear, colorless liquid.

Formula: CH₃COCH₃
MW: 58.08 g/mol
Boiling Pt: 56.2
Melting Pt: -95.4 °C
Density: 0.792 g/cm³ (20 °C)
Flash Pt: <–20 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00008765
CAS Number: 67-64-1
EINECS: 200-662-2
UN: 1090
ADR: 3 II
REACH: 01-2119471330-49
Merck Index: 13,00067
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de5d8e3bf7a94692d684c8f0ccdfb8500fde76d&vid=24fb2e0ec86974b1803780c7d5b23804a085df64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Water (H2O) 0.5% Max.
Residue after Evaporation 0.001% Max.
GC Purity ([CH3]2CO) 99.5% Min.
Solubility in Water Passes Test
Color (APHA) 10 APHA Max.
Titratable Acid (μeq/g) 0.3 μeq/g Max.
Titratable Base (μeq/g) 0.6 μeq/g Max.
Aldehyde (as HCHO) 0.002% Max.
Isopropyl Alcohol ((CH3CHOHCH3) 0.05% Max.
Methanol (CH3OH) 0.05% Max.
Substances Reducing Permanganate Passes Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR