2-Propanol (≥ 50%) ≥99.8% (by GC), VWR Chemicals BDH®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Isopropyl rubbing alcohol, sec-Propanol, Isopropanol, 2-Hydroxypropane, Isopropyl Rubbing Alcohol, 2-Hydroxy propane, IPA, sec-Propyl Alcohol, Dimethyl carbinol, Propane, 2-hydroxy-, Isopropyl alcohol, Propan-2-ol

BDH1174-4LP
BDH11744LPCS 225.24 USD
BDH11744LP
2-Propanol (≥ 50%) ≥99.8% (by GC), VWR Chemicals BDH®
2-Propanol

Laboratory reagent suitable for general laboratory use, as well as for cytology and histology.


  • Tested to USP specifications.

Formula: (CH₃)₂CHOH
MW: 60.1 g/mol
Boiling Pt: 82
Melting Pt: -89
Density: 0.8…0.92 g/cm³ (20 °C)
Flash Pt: 12
Storage Temperature: Ambient
MDL Number: MFCD00011674
CAS Number: 67-63-0
EINECS: 200-661-7
UN: 1987
ADR: 3 II
Merck Index: 13,05228
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ceac541820c6046c770c320cb35e80c1343901a5&vid=4a720b44a08c22b968be82ae24c2b3d604bdd52b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Identification by FTIR Equivalent to standard
Appearance Clear, colorless liquid
Water (K.F.) Less than 0.050 %
Refractive index (20 °C) 1.3772 ± 0.0010
Nonvolatile Residue Less than 5 mg/l (0.0005 g/100 ml)
Assay Greater than 99.8 %
Specific Gravity (25 °C) 0.783 - 0.787
Color (APHA) Less than 10
Acidity Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR