ALCH, (∼20% H₂O) ≥80% Reagent alcohol (denatured with methanol and isopropanol), VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Reagent alcohol

BDH1162-4LP BDH1162-19L
BDH1162-4LPCS 280.32 USD
BDH1162-4LP BDH1162-19L
ALCH, (∼20% H₂O) ≥80% Reagent alcohol (denatured with methanol and isopropanol), VWR Chemicals BDH®
Alcohol

Clear, colorless liquid. Denatured with 3.4–4.0% methanol and 3.6–4.4% 2-propanol. Suitable for cytology and histology.


  • Total alcohol: 80%
  • Ethyl alcohol: 72%
  • Denaturant: 3.4–4.0% methanol and 3.6–4.4% 2-propanol


Ordering information: The purchase of Reagent Alcohol in containers larger than 4 liters requires a Letterhead Permit approval by the United States Treasury Tax and Trade Bureau (TTB).

MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1170
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear, colorless liquid
Water 19.0-21.0%
Residue Less than 0.001% (0.0010 g/118 mL)
GC Purity Ethanol 72.0-73.0%
GC Purity Methanol 3.4-4.0%
GC Purity Isopropyl Alcohol 3.6-4.4%
Color (APHA) Less than 10
Specific gravity @ 20ºC 0.852-0.858
Acidity Less than 0.0025%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR