Ethyl acetate ≥99.5% ACS, VWR Chemicals BDH®

Supplier: VWR International
Danger

Synonyms: Acetic acid ethyl ester, Acetic ether, Acetoxyethane, Acetidin, Ethyl acetic ester, Ethyl ethanoate, EtOAc, Vinegar naphtha, Acetic acid ethyl ester

BDH1123-19L BDH1123-4LP BDH1123-4LG
BDH1123-19LEA 929.77 USD
BDH1123-19L BDH1123-4LP BDH1123-4LG
Ethyl acetate ≥99.5% ACS, VWR Chemicals BDH®
Ethyl acetate

Meets ACS Specifications for General Use

Formula: CH₃COOC₂H₅
Boiling Pt: 77.1 °C (1013 hPa)
Melting Pt: –83 °C
Density: 0.902 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3,II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, colorless liquid
Water (K.F.) Less than 0.20%
Residue Less than 0.003% (0.0012 g/45mL)
GC Purity Greater than 99.5%
Color (APHA) Less than 10
Titrable acid Less than 0.9 meq/g (Not more than 0.80 mL of 0.01N sodium hydroxide)
Substances darkened by sulfuric acid Passes test (no dark color at interface)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR