BD Perisafe™ Thin Wall Epidural Needles, BD Medical
Supplier: BD
405092
BD405092CS 659.65 USD
BD405092
BD Perisafe™ Thin Wall Epidural Needles, BD Medical
Syringe Needles
Needles feature a modified Tuohy point designed to provide enhanced tactile feel
Their short point length minimizes accidental dural puncture. These needles are slightly sharper than tactile needles for penetrating tough tissue, and the needle heel is specially designed to minimize catheter shearing. Calibrated needle depth markings assist in awareness of needle placement without interfering with tactile feedback. A unique, fitted one-piece plastic stylet prevents tissue coring.