Atpenin A5 ≥95%

Supplier: ALADDIN SCIENTIFIC

A275010-1MG A275010-5MG
ALA275010-1MGEA 290.61 USD
ALA275010-1MG ALA275010-5MG
Atpenin A5 ≥95%
Atpenin A5

Store at -20°C. Store under desiccating conditions. The product can be stored for up to 12 months.

Formula: C₁₅H₂₁Cl₂NO₅
Melting Pt: 83-86° C
Density: 1.31 g/cm³
Storage Temperature: Freezer
CAS Number: 119509-24-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76a2ab5e7a4dd92f0516a2899895185714280000&vid=76a2ab5e7a4dd92f0516a2899895185714280000&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR