7-Aza-spiro[3.5]nonane hydrochloride ≥97%

Supplier: ALADDIN SCIENTIFIC
Warning

Synonyms: 7-Azaspiro[3.5]nonane hydrochloride

A174040-100MG A174040-1G A174040-250MG
ALA174040100MGEA 36.96 USD
ALA174040100MG ALA174040-1G ALA174040250MG
7-Aza-spiro[3.5]nonane hydrochloride ≥97%
7-Aza-spiro[3.5]nonane hydrochloride
Formula: C₈H₁₅N·ClH
MW: 161.672 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD23105809
CAS Number: 1414885-16-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=274b34302f61e162e3a37650aa3c1a22acaaa497&vid=274b34302f61e162e3a37650aa3c1a22acaaa497&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR